accessibility menu, dialog, popup

UserName

OFeOFeONiO

CAS RN 12168-54-6

IUPAC Name: diiron(3+) dinickel(2+) pentaoxidandiide
Synonyms: diiron(3+) dinickel(2+) pentaoxidandiide
Molecular Weight (g/mol): 309.07
Molecular Formula: Fe2Ni2O5
InChi Key: JEALIFOJMITGDQ-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Ni++].[Ni++]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results