missing translation for 'onlineSavingsMsg'
Learn More

Iron nickel oxide, tech.

Product Code. 11329289
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329289 50 g 50g
11339289 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329289 Supplier Thermo Scientific Alfa Aesar Supplier No. 088530.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12168-54-6 | Fe2NiO4 | 234.38 g/mol

Iron nickel oxide is used as an adsorbent to remove hexavalent chromium ions present in the waste water by co-sedimentation method. It is also employed as an additive in membrane (i.e. polyvinylchloride/styrene-butadiene-rubber blend heterogeneous cation exchange membrane) fabrication, thereby increasing the permeability for bivalent ions compared to the monovalent type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12168-54-6
Molecular Formula Fe2NiO4
Molecular Weight (g/mol) 234.38
MDL Number MFCD00016254
InChI Key NQNBVCBUOCNRFZ-UHFFFAOYSA-N
IUPAC Name oxo[(oxoferrio)oxy]iron; oxonickel
SMILES O=[Ni].O=[Fe]O[Fe]=O

Specifications

Color Brown
Physical Form Powder
Quantity 50 g
Formula Weight 234.39
Grade Technical
Odor Odorless
Appearance Dark-brown powder
Density 5.368 g/cm3 at 20°C
Chemical Name or Material Iron nickel oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.