missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Irisin/FNDC5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£145.00 - £386.00
Specifications
| Antigen | Irisin/FNDC5 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227483
|
Novus Biologicals
NBP2-59680SS |
0.025 mg |
£145.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18292475
|
Novus Biologicals
NBP2-59680 |
0.1 mg |
£386.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Irisin/FNDC5 Polyclonal specifically detects Irisin/FNDC5 in Human, Mouse, Camel samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Irisin/FNDC5 | |
| Western Blot 0.5 - 1.0 ug/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism, Neuroscience, Stem Cells | |
| PBS with 0.02% Sodium Azide | |
| 252995 | |
| Partial recombinant protein made to an internal region of the human Irisin/FNDC5 protein (amino acids 32-143) [UniProt Q8NAU1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Camelid | |
| fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2 | |
| FNDC5 | |
| IgG | |
| Affinity Purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title