missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Irisin/FNDC5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-59680
This item is not returnable.
View return policy
Description
Irisin/FNDC5 Polyclonal specifically detects Irisin/FNDC5 in Human, Mouse, Camel samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Irisin/FNDC5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FNDC5 | |
| Partial recombinant protein made to an internal region of the human Irisin/FNDC5 protein (amino acids 32-143) [UniProt Q8NAU1] | |
| 0.1 mg | |
| Lipid and Metabolism, Neuroscience, Stem Cells | |
| 252995 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5 - 1.0 ug/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Camelid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction