missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan Lactone Impurity, TRC

Product Code. 30394046
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394046 25 mg
30406836 50 mg
30405772 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394046 Supplier Toronto Research Chemicals Supplier No. TRCI76752025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143490-53-3
Molecular Formula C32 H36 N4 O5
Molecular Weight (g/mol) 556.65
Synonym [1,4'-Bipiperidine]-1'-carboxylic acid, 3,10-diethyl-11,13-dihydro-1,13-dioxo-1H,3H-furo[3',4':6,7]indolizino[1,2-b]quinolin-8-yl ester, [1,4'-Bipiperidine]-1'-carboxylic Acid 3,10-Diethyl-11,13-dihydro-1,13-dioxo-1H,3H- furo[3',4':6,7]indolizino[1,2-b]quinolin-8-yl Ester
SMILES CCC1OC(=O)C2=C1C=C3N(Cc4c(CC)c5cc(OC(=O)N6CCC(CC6)N7CCCCC7)ccc5nc34)C2=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C32H36N4O5/c1-3-21-22-16-20(40-32(39)35-14-10-19(11-15-35)34-12-6-5-7-13-34)8-9-25(22)33-29-24(21)18-36-26(29)17-23-27(4-2)41-31(38)28(23)30(36)37/h8-9,16-17,19,27H,3-7,10-15,18H2,1-2H3
Formula Weight 556.2686
Chemical Name or Material Irinotecan Lactone Impurity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.