CAS RN 143490-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Irinotecan Lactone Impurity, TRC
CAS: 143490-53-3 Molecular Formula: C32 H36 N4 O5 Molecular Weight (g/mol): 556.65 Synonym: [1,4'-Bipiperidine]-1'-carboxylic acid, 3,10-diethyl-11,13-dihydro-1,13-dioxo-1H,3H-furo[3',4':6,7]indolizino[1,2-b]quinolin-8-yl ester,[1,4'-Bipiperidine]-1'-carboxylic Acid 3,10-Diethyl-11,13-dihydro-1,13-dioxo-1H,3H- furo[3',4':6,7]indolizino[1,2-b]quinolin-8-yl Ester SMILES: CCC1OC(=O)C2=C1C=C3N(Cc4c(CC)c5cc(OC(=O)N6CCC(CC6)N7CCCCC7)ccc5nc34)C2=O