Learn More
Iridium black, 99.95% (metals basis), Thermo Scientific™
£387.00 - £1465.00
Chemical Identifiers
| CAS | 7439-88-5 |
|---|---|
| Molecular Formula | Ir |
| Molecular Weight (g/mol) | 192.22 |
| MDL Number | MFCD00011062 |
| InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
| Synonym | ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge |
| PubChem CID | 23924 |
| ChEBI | CHEBI:49666 |
| IUPAC Name | iridium |
| SMILES | [Ir] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15432877
|
Thermo Scientific Alfa Aesar
047150.ME |
500mg |
£387.00
500mg |
Estimated Shipment: 29-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15412877
|
Thermo Scientific Alfa Aesar
047150.04 |
2g |
£1465.00
2g |
Estimated Shipment: 06-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| Ir | |
| MFCD00011062 | |
| ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
| CHEBI:49666 | |
| [Ir] |
Specifications
| 7439-88-5 | |
| MFCD00011062 | |
| ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge | |
| [Ir] | |
| 192.22 | |
| CHEBI:49666 | |
| (metals basis) |
| Ir | |
| UN3089 | |
| GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
| iridium | |
| 23924 | |
| 99.95% | |
| Iridium black |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-095-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.