missing translation for 'onlineSavingsMsg'
Learn More

Iridium black, 99.95% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 047150.04

Quantity 2g

 View more versions of this product

Product Code. 15412877

  • £1465.00 / 2g
 Restricted Item
Estimated Shipment: 06-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7439-88-5
192.22
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
CHEBI:49666
[Ir]

Specifications

7439-88-5
Ir
UN3089
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
99.95%
Iridium black
2g
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
[Ir]
192.22
CHEBI:49666
(metals basis)
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.