missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£189.00 - £380.00
Specifications
| Antigen | IRF9 |
|---|---|
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18333644
|
Novus Biologicals
NBP3-03263-20UL |
20 μL |
£189.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301476
|
Novus Biologicals
NBP3-03263-100UL |
100 μL |
£380.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRF9 Polyclonal specifically detects IRF9 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
| IRF9 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 113-255 of human IRF9 (NP_006075.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 10379 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title