missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03263-100UL
This item is not returnable.
View return policy
Description
IRF9 Polyclonal specifically detects IRF9 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| IRF9 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, KnockDown | |
| Unconjugated | |
| IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 113-255 of human IRF9 (NP_006075.3). | |
| 100 μL | |
| Signal Transduction | |
| 10379 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction