missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF9 Mouse anti-Human, Clone: PCRP-IRF9-2F8, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £423.00
Specifications
| Antigen | IRF9 |
|---|---|
| Clone | PCRP-IRF9-2F8 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328836
|
Novus Biologicals
NBP3-13835-20UG |
20 μg |
£216.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18359386
|
Novus Biologicals
NBP3-13835-100UG |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein ArraySpecifications
| IRF9 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
| Recombinant full-length human IRF9 protein (Uniprot: Q00978) | |
| Primary | |
| Store at 4°C. |
| PCRP-IRF9-2F8 | |
| Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q00978 | |
| 10379 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title