missing translation for 'onlineSavingsMsg'
Learn More

IRF9 Mouse anti-Human, Clone: PCRP-IRF9-2F8, Novus Biologicals™

Product Code. 18328836 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328836 20 μg 20µg
18359386 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18328836 Supplier Novus Biologicals Supplier No. NBP31383520UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF9
Applications Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-IRF9-2F8
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q00978
Gene Alias IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma
Host Species Mouse
Immunogen Recombinant full-length human IRF9 protein (Uniprot: Q00978)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10379
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.