Learn More
Invitrogen™ IRF3 Monoclonal Antibody (SL-12 (SL12)), eBioscience™

Mouse Monoclonal Antibody
£359.00 - £1376.00
Specifications
| Antigen | IRF3 |
|---|---|
| Clone | SL-12 (SL12) |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17190832
|
Invitrogen™
14-9947-37 |
2 mg |
£1376.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15228267
![]() |
Invitrogen™
14-9947-82 |
100 μg |
£359.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This SL-12 monoclonal antibody reacts with human Interferon Regulatory Factor (IRF)-3. IRF-3 is a 427 amino acid constitutively expressed member of the IRF transcription factor family that plays a critical role in the induction of type I interferons. Inactive IRF-3 continuously transits between the cytoplasm and nucleus. Virus or dsRNA induces the C-terminal phosphorylation of IRF-3 by IKKε and TBK-1. This phosphorylation results in retention of IRF-3 in the nucleus and subsequent transcription of IFN-β, several interferon stimulated genes (ISGs), and other antiviral response genes. Applications Reported: This SL-12 (SL12) antibody has been reported for use in immunoblotting (WB). Applications Tested: This SL-12 (SL12) antibody has been tested by immunoblot of normal human peripheral blood cell lysates. This antibody can be used at less than or equal to 2 μg/mL in blocking buffer. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
IFR3 (interferon regulatory factor 3) is a key transcriptional regulator of type I interferon (IFN)-dependent immune responses. It plays a critical role in the innate immune response against DNA and RNA viruses. IFR3 regulates the transcription of type I IFN genes (IFN-alpha and IFN-beta) and IFN-stimulated genes (ISG) by binding to an interferon-stimulated response element (ISRE) in their promoters. It acts as a more potent activator of the IFN-beta gene than the IFN-alpha gene and plays a critical role in both the early and late phases of the IFNA/B gene induction. IFR3 is found in an inactive form in the cytoplasm of uninfected cells and following viral infection, double-stranded RNA, or toll-like receptor signaling. It is phosphorylated by IKBKE and TBK1 kinases. This induces a conformational change, leading to its dimerization and nuclear localization and association with CREB binding protein to form dsRNA-activated factor 1 (DRAF1), a complex which activates the transcription of the type I IFN and ISG genes. Mutations in the gene can result in encephalopathy, acute, infection-induced, herpes-specific 7.Specifications
| IRF3 | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q14653 | |
| 3661 | |
| Primary | |
| Affinity chromatography |
| SL-12 (SL12) | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| IRF3 | |
| C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3 | |
| IRF3 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.