Learn More
Invitrogen™ IRF3 Monoclonal Antibody (SL-12 (SL12)), eBioscience™

Description
Description: This SL-12 monoclonal antibody reacts with human Interferon Regulatory Factor (IRF)-3. IRF-3 is a 427 amino acid constitutively expressed member of the IRF transcription factor family that plays a critical role in the induction of type I interferons. Inactive IRF-3 continuously transits between the cytoplasm and nucleus. Virus or dsRNA induces the C-terminal phosphorylation of IRF-3 by IKKε and TBK-1. This phosphorylation results in retention of IRF-3 in the nucleus and subsequent transcription of IFN-β, several interferon stimulated genes (ISGs), and other antiviral response genes. Applications Reported: This SL-12 (SL12) antibody has been reported for use in immunoblotting (WB). Applications Tested: This SL-12 (SL12) antibody has been tested by immunoblot of normal human peripheral blood cell lysates. This antibody can be used at less than or equal to 2 μg/mL in blocking buffer. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | IRF3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SL-12 (SL12) |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | IRF3 |
| Gene Accession No. | Q14653 |
| Gene Alias | C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.