missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
£168.00
Specifications
| Antigen | IRAK4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:2000, Flow Cytometry, Immunoprecipitation |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IRAK4 Polyclonal specifically detects IRAK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
| IRAK4 | |
| Western Blot 1:1000 - 1:2000, Flow Cytometry, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| NP_001107654 | |
| 51135 | |
| This antibody was developed against a mixture of synthetic peptides containing amino acids 38-54 and 120-136 of mouse IRAK-4. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS with 0.05% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
| IRAK4 | |
| IgG | |
| Protein G purified | |
| Cross-reacts with Human and Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title