missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NB500-597-0.05mg
This item is not returnable.
View return policy
Description
IRAK4 Polyclonal specifically detects IRAK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications
| IRAK4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross-reacts with Human and Mouse. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:2000, Flow Cytometry, Immunoprecipitation | |
| NP_001107654 | |
| IRAK4 | |
| This antibody was developed against a mixture of synthetic peptides containing amino acids 38-54 and 120-136 of mouse IRAK-4. | |
| 0.05 mg | |
| Protein Kinase | |
| 51135 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction