missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£185.00
Specifications
| Antigen | IRAK1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 5 μg/mL, Immunoprecipitation 2-4 μg per sample |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IRAK1 Polyclonal specifically detects IRAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| IRAK1 | |
| Western Blot 1 - 5 μg/mL, Immunoprecipitation 2-4 μg per sample | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P51617 | |
| 3654 | |
| Rabbit anti-IRAK polyclonal antibody was raised against a peptide corresponding to amino acids 700 to 712 of human IRAK. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle | |
| IRAK1 | |
| IgG | |
| Protein G purified | |
| Has no cross reactivity to IRAK2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title