missing translation for 'onlineSavingsMsg'
Learn More

IRAK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-56699SS

 View more versions of this product

Product Code. 18525762

  • £185.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IRAK1 Polyclonal specifically detects IRAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRAK1
Polyclonal
Unconjugated
PBS containing 0.05% BSA with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle
Rabbit
Protein G purified
RUO
Primary
Has no cross reactivity to IRAK2.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Western Blot
0.5 mg/mL
Western Blot 1 - 5 μg/mL, Immunoprecipitation 2-4 μg per sample
P51617
IRAK1
Rabbit anti-IRAK polyclonal antibody was raised against a peptide corresponding to amino acids 700 to 712 of human IRAK.
0.025 mg
Protein Kinase
3654
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only