Learn More
Eppendorf IQ/OQ GxP Services
Product Code. 32215727 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
CellXpert 170(i)
CellXpert CS220
Heating Shaker
Heating/Cooling Shaker
S41i Incubator Shaker
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
32215727 CellXpert CS220 Each
17191056 S41i Incubator Shaker Each
17101066 CellXpert 170(i) Each
17121066 Heating Shaker Each
17131066 Heating/Cooling Shaker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 32215727 Supplier Eppendorf Supplier No. 0082050302
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installation and operational qualification assures that instrument is installed in accordance with Eppendorf's specifications within defined test protocols.
  • Check of delivery status and installation environment
  • Initial configuration, functional (according to manufacturer specifications) and safety checks
  • Detailed IQ report
  • Test of notification and alert functions
  • Verification of instrument parameters (according to manufacturer specifications)
  • Detailed OQ report
  • Handling of deviation list/management
  • Signed approval by customer and service technician
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CellXpert CS220
Product Type IQ/OQ Service