missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ipsapirone
GREENER_CHOICE

Product Code. 30001410
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
30001410 50 mg 50mg
18044482 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 30001410

Brand: Tocris Bioscience™ 1869/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT1A agonist

Selective 5-HT1A receptor agonist (Ki = 10 nM). Anxiolytic in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95847-70-4
Molecular Formula C19H23N5O3S

Specifications

Quantity 50 mg
Formula Weight 401.48
Purity >99%
Molecular Weight (g/mol) 401.485
Chemical Name or Material Ipsapirone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.