accessibility menu, dialog, popup

UserName

CAS RN 95847-70-4

ONNNNNSOO

CAS RN 95847-70-4

IUPAC Name: 2-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Synonyms: ipsapirone
Molecular Weight (g/mol): 401.49
Molecular Formula: C19H23N5O3S
InChi Key: TZJUVVIWVWFLCD-UHFFFAOYSA-N
SMILES: O=C1N(CCCCN2CCN(CC2)C2=NC=CC=N2)S(=O)(=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results