missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Iparomlimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559439
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Iparomlimab, a biosimilar within the category of programmed cell death protein 1 (PD-1) inhibitors. The gene PDCD1, also known as PD-1 or CD279, encodes Programmed Cell Death Protein 1, a crucial immune checkpoint receptor expressed on T cells and other immune cells. Located on chromosome 2q37.3, PDCD1 consists of five exons. PD-1 is a type I transmembrane protein within the CD28/CTLA-4 superfamily and functions primarily to inhibit immune responses, thus maintaining peripheral tolerance and preventing autoimmune reactions. Upon binding to its ligands, PD-L1 and PD-L2, expressed on tumor cells and other tissues, PD-1 mediates the suppression of T-cell activity, inducing apoptosis in antigen-specific T cells while sparing regulatory T cells, contributing significantly to immune evasion by tumors.
Specifications
| Iparomlimab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction