missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ionomycin, 96%

Product Code. p-7070072
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15414829

Brand: Thermo Scientific Chemicals J62448.M^

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly selective and potent calcium ionophore | CAS: 56092-81-0 | C41H72O9, C41H72O9 | 709.02 g/mol

Ionomycin free acid is a beneficial tool in research to raise intracellular calcium levels. Also this antibiotic can be used to study the calcium transport across the plasma membrane. It has been shown to induce central demyelination , inhibit adrenal bovine TREK-1 channels , and to regulate cell division of mature human B cells . It is used to study the effects of calcium flux on endoplasmic reticulum (ER) stress, mitochondrial stress and intrinsic apoptosis mechanisms. It is also used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2, and IL-4 usually in conjunction with PMA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-81-0
Molecular Formula C41H72O9

Specifications

Quantity 1 mg
Description Colorless
Content And Storage -30°C to -10°C
For Use With (Application) Highly selective and potent calcium ionophore
Formula Weight 709.02
Odor Alcohol-like
Product Type Ionomycin
Form Solution in ethanol
Beilstein 3642126
InChI Key PGHMRUGBZOYCAA-OJFQOPKISA-N
SMILES C[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)CC1CC[C@](C)(O1)C1CC[C@](C)(O1)[C@@H](C)O
IUPAC Name (4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-trihydroxy-22-[(2S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9-oxodocosa-10,16-dienoic acid
Molecular Weight (g/mol) 709.02
Assay 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ionomycin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, ionomycin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.