Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ TOP10F' Chemically Competent E. coli, no manual
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
Brand: Invitrogen™ KNM455001
Code : 50
Additional Details : Weight : 0.00500kg
Description
Uses a pCR™-TOPO™ Vector with covalently bound topoisomerase I for fast cloning and recombinants.
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- Choice of T7 (pCR2.1-TOPO) or T7 and SP6 (pCRII-TOPO) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
- EcoR I sites flanking the PCR product insertion site for easy excision of inserts
- Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
- Easy blue/white colony screening for selection of recombinants
- TOPO TA Cloning Kits are also available in combo kits combined with a PureLink™Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO-cloned inserts for downstream analysis
Cloning, PCR Cloning
Specifications
Kit, Cloning | |
TOP10F´ | |
25 Reactions |
Box 1: Topoisomerase I-activated pCR2.1-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid | |
Cloning, PCR Cloning |
For Research Use Only. Not for use in diagnostic procedures.