missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ MAX Efficiency™ DH5α-T1R E. coli

Designed for cloning PCR products directly from a PCR reaction in just 5 minutes

Brand:  Invitrogen™ K452040

Code : 50

Additional Details : Weight : 0.12000kg

 View more versions of this product

Product Code. 10462862

  • £1012.00 / Each
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

They use a pCR™-TOPO Vector with covalently bound topoisomerase I for fast cloning and recombinants.

  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • Choice of T7 (pCR2.1-TOPO) or T7 and SP6 (pCRII-TOPO) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
  • EcoR I sites flanking the PCR product insertion site for easy excision of inserts
  • Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants

Cloning, PCR Cloning

Specifications

Specifications

DH5α
Box 1:
• Topoisomerase I-activated pCR™2.1-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.
One Shot™
T7, SP6
TOPO-TA Cloning Vectors
TOPO™-TA
Kit
Cloning Kit
50 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.