missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ MAX Efficiency™ DH5α-T1R E. coli
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
£638.00 - £1012.00
Specifications
Bacterial or Yeast Strain | DH5α |
---|---|
Cloning Method | TOPO™-TA |
Content And Storage | Box 1: • Topoisomerase I-activated pCR™2.1-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid Store at -68 to -85°C. |
Format | Kit |
Product Line | One Shot™ |
Includes: Topoisomerase I-activated pCR™2.1-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10624043
|
Invitrogen™
K452001 |
25 Reactions |
£638.00
Each |
Estimated Shipment: 08-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10462862
|
Invitrogen™
K452040 |
50 Reactions |
£1012.00
Each |
Estimated Shipment: 08-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
They use a pCR™-TOPO Vector with covalently bound topoisomerase I for fast cloning and recombinants.
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- Choice of T7 (pCR2.1-TOPO) or T7 and SP6 (pCRII-TOPO) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
- EcoR I sites flanking the PCR product insertion site for easy excision of inserts
- Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
- Easy blue/white colony screening for selection of recombinants
Cloning, PCR Cloning
Specifications
DH5α | |
Box 1: • Topoisomerase I-activated pCR™2.1-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid Store at -68 to -85°C. |
|
One Shot™ | |
T7, SP6 |
TOPO™-TA | |
Kit | |
Cloning Kit | |
TOPO-TA Cloning Vectors |
For Research Use Only. Not for use in diagnostic procedures.