missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Pan Mouse IgG

Dynabeads™ Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion.

Brand:  Invitrogen™ 11042

1284.06 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 10680894

  • £1926.00 / Each
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes: 5mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
This item is not returnable. View return policy

Description

Description

Dynabeads™ Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

Depletion—cells can be depleted from a sample
Positive isolation—cells can be positively isolated from a sample for
molecular applications
Negative isolation—cells can be negatively isolated from a PBMC sample by
adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible
with flow cytometry.

Note:
For cellular assays we recommend positive isolation followed by release of beads from the cells. Recommended product for bead-free cells: Dynal™ CELLection™ Pan Mouse IgG (Cat. No. 115-31D) in combination with your mouse IgG.

Starting samples:
Whole blood, PBMC, buffy coat, tissue digests.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Whole Antibody
Monoclonal
Antibody Coated Bead
Human
Processes ∼2 x 109 cells total
DYNAL™, Dynabeads™
5 x 5 mL
PBMC, Tissue Digests, Blood
1 x 107 PBMCs per isolation
All cells from all species, except mouse
This Product Contains: 5 x 5 mL Dynabeads™ Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is a unique Fc-specific human* anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
Store at 2°C to 8°C.
Dynabeads™ Pan
Depletion, negative isolation or positive isolation
>95%
Research Grade
All species, except mouse
Room Temperature
All species, except mouse
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.