Learn More
Invitrogen™ Dynabeads™ Pan Mouse IgG
Dynabeads™ Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion.
£572.00 - £1926.00
Specifications
Antibody Fragment | Whole Antibody |
---|---|
Cell Type | All cells from all species, except mouse |
Clonality | Monoclonal |
Product Type | Antibody Coated Bead |
Dye Type | Dynabeads™ Pan |
Includes: 5mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10542795
|
Invitrogen™
11041 |
5 mL |
£572.00
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
10680894
|
Invitrogen™
11042 |
5 x 5 mL |
£1926.00
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
Dynabeads™ Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.
• Depletion—cells can be depleted from a sample
• Positive isolation—cells can be positively isolated from a sample for
molecular applications
• Negative isolation—cells can be negatively isolated from a PBMC sample by
adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible
with flow cytometry.
Note:
For cellular assays we recommend positive isolation followed by release of beads from the cells. Recommended product for bead-free cells: Dynal™ CELLection™ Pan Mouse IgG (Cat. No. 115-31D) in combination with your mouse IgG.
Starting samples:
Whole blood, PBMC, buffy coat, tissue digests.
Order Info
Shipping Condition: Room Temperature
Specifications
Whole Antibody | |
Monoclonal | |
Dynabeads™ Pan | |
Depletion, negative isolation or positive isolation | |
>95% | |
Research Grade | |
PBMC, Tissue Digests, Blood | |
1 x 107 PBMCs per isolation |
All cells from all species, except mouse | |
Antibody Coated Bead | |
Human | |
Processes ∼2 x 109 cells total | |
DYNAL™, Dynabeads™ | |
All species, except mouse | |
Room Temperature | |
All species, except mouse |
For Research Use Only. Not for use in diagnostic procedures.