missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS5 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | INTS5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18353205
|
Novus Biologicals
NBP3-04514-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18335976
|
Novus Biologicals
NBP3-04514-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INTS5 Polyclonal specifically detects INTS5 in Human, Mouse samples. It is validated for Western Blot.Specifications
| INTS5 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 80789 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Int5, integrator complex subunit 5, KIAA1698INT5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-400 of human INTS5 (NP_085131.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel