missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intron-Binding Protein Of 160 KDa Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£196.00 - £558.00
Specifications
| Antigen | Intron-Binding Protein Of 160 KDa |
|---|---|
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18102955
|
Novus Biologicals
NBP2-23607-0.025MG |
0.025 mg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152384
|
Novus Biologicals
NBP2-23607-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18178721
|
Novus Biologicals
NBP2-23607 |
0.1 mg |
£558.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Intron-Binding Protein Of 160 KDa Polyclonal specifically detects Intron-Binding Protein Of 160 KDa in Human, Mouse, Rat samples. It is validated for Western Blot.Tekniske data
| Intron-Binding Protein Of 160 KDa | |
| Western Blot | |
| Unconjugated | |
| O60306 | |
| 9716 | |
| Synthetic peptide corresponding to the following peptide sequence: DYILLSLVRTRAVGHLC | |
| Primary |
| Western Blot 1:500 | |
| Polyclonal | |
| Rabbit | |
| AQR, aquarius homolog (mouse), FSAP164, Functional Spliceosome-Associated Protein 164, IBP160, Intron-Binding Protein Aquarius | |
| AQR | |
| IgG |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel