missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intron-Binding Protein Of 160 KDa Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23607-0.025MG
This item is not returnable.
View return policy
Description
Intron-Binding Protein Of 160 KDa Polyclonal specifically detects Intron-Binding Protein Of 160 KDa in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Intron-Binding Protein Of 160 KDa | |
| Polyclonal | |
| Western Blot 1:500 | |
| O60306 | |
| AQR | |
| Synthetic peptide corresponding to the following peptide sequence: DYILLSLVRTRAVGHLC | |
| 0.025 mg | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| AQR, aquarius homolog (mouse), FSAP164, Functional Spliceosome-Associated Protein 164, IBP160, Intron-Binding Protein Aquarius | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9716 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction