missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (2G18) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£158.00 - £426.00
Specifications
| Antigen | Integrin beta 7 |
|---|---|
| Clone | 2G18 |
| Concentration | 0.4 mg/mL |
| Dilution | Flow Cytometry 1:50-200, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:100-400, Immunohistochemistry-Paraffin 1:100-200 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18259892
|
Novus Biologicals
NB110-94147-100UG |
100 μg |
£426.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18074466
|
Novus Biologicals
NB110-94147-0.025MG |
0.025 mg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Integrin beta 7 | |
| 0.4 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| ITGB7 | |
| IgG2 | |
| Protein G purified | |
| This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. |
| 2G18 | |
| Flow Cytometry 1:50-200, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:100-400, Immunohistochemistry-Paraffin 1:100-200 | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| 3695 | |
| Cell lysates of mouse lymphocytes | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title