missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 7 Antibody (2G18) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-94147-100UG
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Integrin beta 7 | |
| Monoclonal | |
| 0.4 mg/mL | |
| Flow Cytometry 1:50-200, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:100-400, Immunohistochemistry-Paraffin 1:100-200 | |
| Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 | |
| Rat | |
| Protein G purified | |
| Signal Transduction | |
| 3695 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 2G18 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| ITGB7 | |
| Cell lysates of mouse lymphocytes | |
| 100 μg | |
| Primary | |
| This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction