missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen Integrin beta-3
Concentration 0.27 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898384
View Documents
Proteintech
18309-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888384
View Documents
Proteintech
18309-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ITGB3 is involved in Glanzmann thrombasthenia (GT), a common inherited disease of platelet aggregation. It is characterized by mucocutaneous bleeding of mild-to-moderate severity. GT has been classified clinically into types I and II. In type I, platelets show absence of the glycoprotein IIb-IIIa complexes at their surface and lack fibrinogen and clot retraction capability. In type II, the platelets express the GPIIb-IIIa complex at reduced levels, have detectable amounts of fibrinogen, and have low or moderate clot retraction capability. The disease is caused by mutations affecting the gene represented in this entry.
Specifications

Specifications

Integrin beta-3
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Rat, Human
O54890, P05106
16416, 29302, 3690
Peptide
Primary
-20°C
ITGB3
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CD61, GP3A, GPIIIa, Integrin beta 3, Integrin beta-3, ITGB3
ITGB3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documenti

Documenti

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato