missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18309-1-AP-150UL
This item is not returnable.
View return policy
Description
ITGB3 is involved in Glanzmann thrombasthenia (GT), a common inherited disease of platelet aggregation. It is characterized by mucocutaneous bleeding of mild-to-moderate severity. GT has been classified clinically into types I and II. In type I, platelets show absence of the glycoprotein IIb-IIIa complexes at their surface and lack fibrinogen and clot retraction capability. In type II, the platelets express the GPIIb-IIIa complex at reduced levels, have detectable amounts of fibrinogen, and have low or moderate clot retraction capability. The disease is caused by mutations affecting the gene represented in this entry.Specifications
| Integrin beta-3 | |
| Polyclonal | |
| Unconjugated | |
| ITGB3 | |
| CD61, GP3A, GPIIIa, Integrin beta 3, Integrin beta-3, ITGB3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16416, 29302, 3690 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54890, P05106 | |
| ITGB3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction