missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 6 Antibody (Av beta6 62ow.17) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£169.00 - £319.00
Specifications
| Antigen | Integrin alpha V beta 6 |
|---|---|
| Clone | Av beta6 62ow.17 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607328
|
Novus Biologicals
NBP2-50450-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618287
|
Novus Biologicals
NBP2-50450 |
0.1 mL |
£319.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human, Mouse (Negative) samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Integrin alpha V beta 6 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| PBS | |
| 3685 | |
| IgG1 | |
| Protein G purified |
| Av beta6 62ow.17 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse (Negative) | |
| CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
| Mouse fibroblasts retrovirally infected with cDNA to human beta6 (3T3 beta6.19). Cells expressed a mouse/human av beta6 chimera. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title