Learn More
Integrin alpha V beta 6 Antibody (Av beta6 62ow.17) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50450
 View more versions of this product
Product Code. 18618287
  • £319.00 / 0.10mL
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha V beta 6
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Mouse fibroblasts retrovirally infected with cDNA to human beta6 (3T3 beta6.19). Cells expressed a mouse/human av beta6 chimera.
0.1 mL
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
3685
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunohistochemistry
Av beta6 62ow.17
Unconjugated
PBS
Rat
Protein G purified
RUO
Primary
Human, Mouse (Negative)
Purified