missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha D/CD11d Antibody (CA11.8H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £356.00
Specifications
| Antigen | Integrin alpha D/CD11d |
|---|---|
| Clone | CA11.8H2 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18032856
|
Novus Biologicals
NB100-65292-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18062126
|
Novus Biologicals
NB100-65292-0.1MG |
0.1 mg |
£356.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha D/CD11d Monoclonal specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Integrin alpha D/CD11d | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| 3681 | |
| Affinity purified CD11 antigen from canine spleen | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CA11.8H2 | |
| Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Canine | |
| ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D | |
| ITGAD | |
| IgG1 | |
| Protein A purified | |
| NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title