missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha D/CD11d Antibody (CA11.8H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65292-0.1MG
This item is not returnable.
View return policy
Description
Integrin alpha D/CD11d Monoclonal specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Integrin alpha D/CD11d | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| CA11.8H2 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| ITGAD | |
| Affinity purified CD11 antigen from canine spleen | |
| 0.1 mg | |
| Signal Transduction | |
| 3681 | |
| Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction