missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (CL7318), Novus Biologicals™
Mouse Monoclonal Antibody
£240.00 - £450.00
Specifications
| Antigen | Integrin alpha 2/CD49b |
|---|---|
| Clone | CL7318 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652171
|
Novus Biologicals
NBP2-76483-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18398520
|
Novus Biologicals
NBP2-76483 |
100 μL |
£450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| Purified | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 3673.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ALEAYSETAKVFSIPFHKDCGEDGLCISDLVLDVRQIPAAQEQPFIVSNQNKRLTFSVTLKNKRESAYNTGIVVDFSENLFFASFSLPV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL7318 | |
| Unconjugated | |
| Mouse | |
| Human | |
| alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
| ITGA2 | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title