Learn More
Invitrogen™ Intact Virus Precipitation Reagent
Description
Intact Virus Precipitation Reagent enables fast and efficient enrichment of intact, live viruses from dilute cell culture and virus transport media. The enrichment is gentle and fast and can be performed in less than three hours. By tying up water molecules, the reagent forces less-soluble components such as vesicles out of solution, allowing them to be collected by short, low-speed centrifugation.
Intact Virus Precipitation Reagent can enrich for a plethora of viruses such as, but not limited to, SARS-CoV-2 coronavirus, respiratory syncytial virus (RSV), and Ebola-, Zika-, influenza A- viruses. The isolated intact virus can be used for functional studies.
Features of Intact Virus Precipitation Reagent include:
• Simple—3-step enrichment requiring only a centrifuge
• Fast—enrichment in <3 hours with minimal hands-on time
• Flexible volume—dilute virus samples can be enriched from large sample volumes
• Straight to PCR—no further nucleic acid extraction needed prior to PCR
Downstream applications
Enriched intact virus can be used for further virus growth or in any downstream functional assays, protein research (western blot), purification of RNA for PCR, etc. Virus isolated using the Intact Virus Precipitation Reagent requires no further nucleic acid extraction prior to doing PCR.
In addition, virus-like particles (VLPs), containing only virus-specific proteins and not nucleic acids, are extensively used in viral proteomic research. Intact Virus Precipitation Reagent isolates both live and inactivated intact viruses and VLPs.
Easy and efficient protocol
The Intact Virus Precipitation Reagent is simply added to the virus sample and incubate for two hours followed by a 30-minute centrifugation step. Remove the supernatant and keep the pellet with the enriched virus that is ready for any downstream analysis.
Automation ready with the KingFisher Purification System
For higher throughput automation needs, use the magnetic bead-based Dynabeads Intact Virus Enrichment (Cat. No. 10700D) strong anion exchange beads using any of our KingFisher Sample Purification Systems or liquid handlers.
Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Intact Virus Precipitation Reagent in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM page.
Specifications
Specifications
| Product Type | Intact Virus Precipitation Reagent |
| Purification Target | Viruses |
| Content And Storage | Store at 2°C to 8°C. |
| For Use With (Equipment) | Centrifuge capable of running upto 10,000 x G |
| Shipping Condition | Ambient Temperature |
| Isolation Technology | Precipitation by centrifugation |
| For Use With (Application) | Precipitation of viruses by tying up water molecules in the sample |
| Quantity | 100 reactions |
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.