missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin/Proinsulin Antibody (D3E7cc), Novus Biologicals™
Mouse Monoclonal Antibody
£93.00 - £199.00
Specifications
| Antigen | Insulin/Proinsulin |
|---|---|
| Clone | D3E7 |
| Dilution | Immunohistochemistry, Enzyme Immunoassay |
| Applications | Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18211860
|
Novus Biologicals
NB200-435SS |
0.04 mg |
£93.00
0.04mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736763
|
Novus Biologicals
NB200-435 |
0.2 mg |
£199.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin/Proinsulin Monoclonal specifically detects Insulin/Proinsulin in Human, Rat, Porcine, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA.Specifications
| Insulin/Proinsulin | |
| Immunohistochemistry, Enzyme Immunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Pig, Bovine | |
| 3630 | |
| Kd for this is 6.3 x 10^8M.This is specific for both insulin and proinsulin | |
| Primary | |
| Store at 4C. Do not freeze. |
| D3E7 | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Diabetes Research | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| INS | |
| IgG1 κ | |
| Protein A purified | |
| This is specific for both insulin and proinsulin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title