missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin/Proinsulin Antibody (D3E7cc), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-435SS
This item is not returnable.
View return policy
Description
Insulin/Proinsulin Monoclonal specifically detects Insulin/Proinsulin in Human, Rat, Porcine, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA.
Specifications
| Insulin/Proinsulin | |
| Monoclonal | |
| Unconjugated | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This is specific for both insulin and proinsulin | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, ELISA | |
| D3E7 | |
| Immunohistochemistry, Enzyme Immunoassay | |
| INS | |
| Kd for this is 6.3 x 10^8M.This is specific for both insulin and proinsulin | |
| 0.04 mg | |
| Diabetes Research | |
| 3630 | |
| Human, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction