missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (IRDN/794), Novus Biologicals™
Mouse Monoclonal Antibody
£231.00 - £391.00
Specifications
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml |
|---|---|
| Classification | Monoclonal |
| Form | Purified |
| Target Species | Pig |
| Gene Alias | IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18780454
|
Novus Biologicals
NBP2-45108-0.02MG |
£231.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18103578
|
Novus Biologicals
NBP2-45108-0.1MG |
0.1 mg |
£391.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
| Purified | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| IgG1 κ | |
| 6 kDa |
| Monoclonal | |
| Pig | |
| INS | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title