missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (IRDN/794), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45108-0.1MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Insulin | |
| Monoclonal | |
| 0.2mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
| P01308 | |
| INS | |
| Recombinant INS protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker. | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| IRDN/794 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| 6 kDa | |
| 0.1 mg | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Human, Mouse, Rat, Pig, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction