missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Instruments Tray

Bochem™ Instruments Tray features rounded corners and includes a lid.

Brand:  Bochem™ 8406

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10040891

  • £204.00 / Each
Estimated Shipment: 12-09-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Stainless Steel
  • Rounded edges
  • Lid included
Specifications

Specifications

250 x 150 x 60 mm
250 mm
Instrument Tray
Lid
60 mm
Stainless Steel
150 mm
Rectangular
Videos
SDS
Documents
Certificates
Special Offers

Special Offers