missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Instruments Accessories - Holder
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
12-Well, Disposable Reagent Reservoirs
24 Eppendorf 1.5 mL and 2.0 mL Tubes
Cluster Tubes, Bullet Tubes or Deep-Well Microplates
Unit Size:
Each
Description
Holder for use with Agilent BioTek Microplate Washers and Readers
Specifications
Specifications
| Product Type | Holder |
| For Use With (Equipment) | 24 Eppendorf 1.5 mL and 2.0 mL Tubes |
| For Use With (Application) | For 24 Eppendorf 1.5 mL and 2.0 mL Tubes |
| No. of Positions | 24 |
| Quantity | 1 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction