Learn More
Invitrogen™ iNOS Recombinant Rat Monoclonal Antibody (CXNFT), Invitrogen™
Rat Recombinant Monoclonal Antibody
£97.75 - £270.00
Specifications
| Antigen | iNOS |
|---|---|
| Clone | CXNFT |
| Concentration | 1.0 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30131582
|
Invitrogen™
740025T20UG |
20 μL |
£97.75
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30131359
|
Invitrogen™
740025T |
100 μL |
£270.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant Rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
iNOS Recombinant Monoclonal antibody specifically detects iNOS in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| iNOS | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| CXNFT | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P29477 | |
| 18126 | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Nos2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.