missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat (R) Gold - 20nm Midi Maleimide Antibody Labeling Kit

Product Code. 18066774 Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Reaction
3 Reactions
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18066774

Brand: Novus Biologicals™ 2710015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

20nm malemide nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • Allows thiolated molecules to be covalently attached to nanoparticles
  • Shields biomolecule from metal surface thus eliminating metal-antibody interactions
  • Reduces risk of sub-optimal antibody performance

Applications: Electron microscopy visualization
Storage: Store at 4°C

Note: Kits are guaranteed for 6 months from date of receipt

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg
Product Type 20nm Mini Maleimide Antibody Labeling Kit
Sufficient For 1 Reaction
Content And Storage -20°C

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.