missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat™ GOLD Antibody Labeling Kit, 80nm, Mini

Product Code. 18397777 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
3 Reactions
Unit Size:
3 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397777 3 Reactions 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397777 Supplier Novus Biologicals™ Supplier No. 23100053Reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unique coated nanoparticle with a strongly bound protective surface coat

TRUSTED_SUSTAINABILITY

Specifications

Description InnovaCoat GOLD 80 nm Mini
Product Type Antibody Labeling Kit
Quantity 3 Reactions
Content And Storage Store at 4°C. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.